(R)-3-Benzyl-1-(tert-butoxycarbonyl)piperidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A246589-10G A246589-25G
77098-608EA 46.68 USD
77098-608 77593-566
(R)-3-Benzyl-1-(tert-butoxycarbonyl)piperidine-3-carboxylic acid 97%
(R)-3-Benzyl-1-(tert-butoxycarbonyl)piperidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₅NO₄
MW: 319.40 g/mol
MDL Number: MFCD26960973
CAS Number: 339539-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR