2',5'-Difluoroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2,5-Difluorophenyl)ethanone, 1-(2,5-Difluorophenyl)ethan-1-one

A246296-25G A246296-500G A246296-5G A246296-100G A246296-10G
77098-596EA 27.59 USD
77098-596 77098-598 77098-600 77685-693 77685-694
2',5'-Difluoroacetophenone 97%
2',5'-Difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆F₂O
MW: 156.13 g/mol
MDL Number: MFCD00009898
CAS Number: 1979-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR