BI-847325 99+%

Supplier: Ambeed
Warning

Synonyms: (Z)-3-(3-((4-((dimethylamino)methyl)phenylamino)(phenyl)methylene)-2-oxoindolin-6-yl)-N-ethylpropiolamide, 3-(3-(((4-((Dimethylamino)methyl)phenyl)amino)(phenyl)methylene)-2-oxoindolin-6-yl)-N-ethylpropiolamide

A245078-1MG A245078-5MG A245078-50MG
77098-584EA 11 USD
77098-584 77098-586 77901-110
BI-847325 99+%
BI-847325

Precaución: for Research Use Only

Formula: C₂₉H₂₈N₄O₂
MDL Number: MFCD28978743
CAS Number: 1207293-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR