1,3-Dichloro-4-iodobenzene 98%

Supplier: Ambeed
Warning

A245050-100G A245050-1G A245050-25G A245050-500G A245050-5G
77098-574EA 82 USD
77098-574 77098-576 77098-578 77098-580 77098-582
1,3-Dichloro-4-iodobenzene 98%
1,3-Dichloro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃Cl₂I
MDL Number: MFCD00001035
CAS Number: 29898-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR