tert-Butyl 4-amino-1H-pyrazole-1-carboxylate 95%

Supplier: Ambeed
Warning

A243534-100MG A243534-1G A243534-250MG A243534-5G
77098-524EA 27.59 USD
77098-524 77098-526 77098-528 77098-530
tert-Butyl 4-amino-1H-pyrazole-1-carboxylate 95%
tert-Butyl 4-amino-1H-pyrazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₃N₃O₂
MW: 183.20 g/mol
MDL Number: MFCD20697038
CAS Number: 1018446-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR