(1R,3S,4S)-tert-Butyl 3-(6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazol-2-yl)-2-azabicyclo[2.2.1]heptane-2-carboxylate 95%

Supplier: Ambeed
Warning

A242722-1G A242722-250MG A242722-5G
77098-500EA 60.48 USD
77098-500 77098-502 77098-504
(1R,3S,4S)-tert-Butyl 3-(6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazol-2-yl)-2-azabicyclo[2.2.1]heptane-2-carboxylate 95%
(1R,3S,4S)-tert-Butyl 3-(6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazol-2-yl)-2-azabicyclo[2.2.1]heptane-2-carboxylate

Precaución: for Research Use Only

Formula: C₂₄H₃₄BN₃O₄
MW: 439.36 g/mol
MDL Number: MFCD28386938
CAS Number: 1256387-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR