3-Chloro-6-(trifluoromethyl)pyridazine 98%

Supplier: Ambeed
Danger

A241404-250MG A241404-1G A241404-25G
77098-476EA 9 USD
77098-476 77612-342 77901-312
3-Chloro-6-(trifluoromethyl)pyridazine 98%
3-Chloro-6-(trifluoromethyl)pyridazine

Precaución: for Research Use Only

Formula: C₅H₂ClF₃N₂
MDL Number: MFCD08275188
CAS Number: 258506-68-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR