(1alpha,3R,4alpha,5R)-4-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-1,3,5-trihydroxycyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

A240499-10MG A240499-1MG A240499-5MG
77098-464EA 69 USD
77098-464 77098-466 77098-468
(1alpha,3R,4alpha,5R)-4-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-1,3,5-trihydroxycyclohexanecarboxylic acid 98%
(1alpha,3R,4alpha,5R)-4-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-1,3,5-trihydroxycyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₈O₉
MDL Number: MFCD10566638
CAS Number: 905-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR