1-(3-Methoxypropyl)piperidin-4-ol 98+%

Supplier: Ambeed
Warning

A240310-1G A240310-25G A240310-5G
77098-456EA 19.1 USD
77098-456 77098-458 77098-460
1-(3-Methoxypropyl)piperidin-4-ol 98+%
1-(3-Methoxypropyl)piperidin-4-ol

Precaución: for Research Use Only

Formula: C₉H₁₉NO₂
MW: 173.25 g/mol
MDL Number: MFCD16153840
CAS Number: 1225518-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR