3-Fluoro-2,4-dimethoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-2,4-dimethoxyphenyl)boronic acid

A238587-1G A238587-250MG A238587-5G
77098-412EA 102.91 USD
77098-412 77098-414 77098-416
3-Fluoro-2,4-dimethoxyphenylboronic acid 98%
3-Fluoro-2,4-dimethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₄
MW: 199.972 g/mol
MDL Number: MFCD23381042
CAS Number: 949146-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR