4,5-Difluoro-2-hydroxyphenylboronic acid 96%

Supplier: Ambeed
Warning

A238019-100MG A238019-1G A238019-250MG
77098-394EA 37.14 USD
77098-394 77098-396 77098-398
4,5-Difluoro-2-hydroxyphenylboronic acid 96%
4,5-Difluoro-2-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₂O₃
MW: 173.91 g/mol
MDL Number: MFCD18914500
CAS Number: 1432610-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR