3-(tert-Butyldimethylsilyloxy)-4-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(t-Butyldimethylsilyloxy)-4-methoxyphenylboronic acid

A237642-1G A237642-250MG A237642-5G
77098-388EA 105.03 USD
77098-388 77098-390 77098-392
3-(tert-Butyldimethylsilyloxy)-4-methoxyphenylboronic acid 98%
3-(tert-Butyldimethylsilyloxy)-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₃BO₄Si
MW: 282.2200 g/mol
MDL Number: MFCD11855851
CAS Number: 900152-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR