(S)-4-(tert-Butoxycarbonyl)-1,4-oxazepane-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A237507-100MG A237507-1G A237507-250MG
77098-374EA 199.45 USD
77098-374 77098-376 77098-378
(S)-4-(tert-Butoxycarbonyl)-1,4-oxazepane-2-carboxylic acid 95%
(S)-4-(tert-Butoxycarbonyl)-1,4-oxazepane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
MW: 245.27 g/mol
MDL Number: MFCD19442252
CAS Number: 1273567-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR