N,N-Diethyl-N-methyl-2-((4-(2-(octyloxy)benzamido)benzoyl)oxy)ethanaminium bromide 98%

Supplier: Ambeed
Warning

Synonyms: Otilonium bromide

A237499-50MG A237499-10MG
77098-372EA 5.31 USD
77098-372 77685-661
N,N-Diethyl-N-methyl-2-((4-(2-(octyloxy)benzamido)benzoyl)oxy)ethanaminium bromide 98%
N,N-Diethyl-N-methyl-2-((4-(2-(octyloxy)benzamido)benzoyl)oxy)ethanaminium bromide

Precaución: for Research Use Only

Formula: C₂₉H₄₃BrN₂O₄
MW: 563.57 g/mol
MDL Number: MFCD00211142
CAS Number: 26095-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR