tert-Butyl 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydrobenzo[f][1,4]oxazepine-4(5H)-carboxylate 95%

Supplier: Ambeed
Warning

A236959-250MG A236959-50MG A236959-100MG
77098-358EA 149 USD
77098-358 77098-360 77685-659
tert-Butyl 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydrobenzo[f][1,4]oxazepine-4(5H)-carboxylate 95%
tert-Butyl 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydrobenzo[f][1,4]oxazepine-4(5H)-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₃₀BNO₅
MW: 375.27 g/mol
MDL Number: MFCD14582651
CAS Number: 1256784-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR