(2S,3R)-3,4-Dihydro-3-isopropyl-2-phenyl-2H-pyrimido[2,1-b]benzothiazole 98%

Supplier: Ambeed
Warning

Synonyms: HBTM-2.1

A236265-100MG A236265-250MG
77098-354EA 55 USD
77098-354 77568-284
(2S,3R)-3,4-Dihydro-3-isopropyl-2-phenyl-2H-pyrimido[2,1-b]benzothiazole 98%
(2S,3R)-3,4-Dihydro-3-isopropyl-2-phenyl-2H-pyrimido[2,1-b]benzothiazole

Precaución: For research use only.

Formula: C₁₉H₂₀N₂S
MW: 308.44 g/mol
MDL Number: MFCD28411847
CAS Number: 1203507-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR