1-Benzyl-3-piperidone hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Benzylpiperidin-3-one hydrochloride

A236162-10G A236162-1G A236162-250MG A236162-5G
77098-346EA 53.05 USD
77098-346 77098-348 77098-350 77098-352
1-Benzyl-3-piperidone hydrochloride 97%
1-Benzyl-3-piperidone hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO·HCl
MW: 225.72 g/mol
MDL Number: MFCD00012791
CAS Number: 50606-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR