3-Methoxy-1-methyl-4-nitro-1H-pyrazole 95%
Supplier: Ambeed
|
Warning
|
A235351-100MG
A235351-1G
A235351-250MG
A235351-5G
77098-338EA
40.32
USD
77098-338
77098-340
77098-342
77098-344
3-Methoxy-1-methyl-4-nitro-1H-pyrazole 95%
3-Methoxy-1-methyl-4-nitro-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₅H₇N₃O₃ MW: 157.12 g/mol |
MDL Number:
MFCD25371334 CAS Number: 1201935-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...