Methyl 2-bromo-3-chlorobenzoate 95%

Supplier: Ambeed
Warning

A235087-1G A235087-5G
77098-334EA 28 USD
77098-334 77098-336
Methyl 2-bromo-3-chlorobenzoate 95%
Methyl 2-bromo-3-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MDL Number: MFCD18398719
CAS Number: 1107627-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR