7-Chloro-3-iodo-1H-pyrrolo[3,2-b]pyridine 95%

Supplier: Ambeed
Warning

A235003-100MG A235003-250MG A235003-50MG
77098-328EA 118.83 USD
77098-328 77098-330 77098-332
7-Chloro-3-iodo-1H-pyrrolo[3,2-b]pyridine 95%
7-Chloro-3-iodo-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄ClIN₂
MW: 278.48 g/mol
MDL Number: MFCD12963113
CAS Number: 1190318-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR