N-Boc-DL-phenylglycine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-DL-Phg-OH

A234866-25G A234866-100G A234866-5G A234866-500G
77098-326EA 23.34 USD
77098-326 77612-298 77612-300 77685-649
N-Boc-DL-phenylglycine 98%
N-Boc-DL-phenylglycine

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄
MW: 251.282 g/mol
MDL Number: MFCD00971063
CAS Number: 3601-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR