3-Isopropyloxetan-3-ol 95%

Supplier: Ambeed
Warning

A234545-100MG A234545-1G A234545-250MG A234545-5G
77098-318EA 61 USD
77098-318 77098-320 77098-322 77098-324
3-Isopropyloxetan-3-ol 95%
3-Isopropyloxetan-3-ol

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MDL Number: MFCD22690227
CAS Number: 1408291-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR