(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-ureidohexanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Homocit-OH, Fmoc-D-HoCit-OH

A233982-10G A233982-5G
77098-310EA 168 USD
77098-310 77098-312
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-ureidohexanoic acid 97%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-ureidohexanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₅N₃O₅
MDL Number: MFCD00237390
CAS Number: 201485-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR