2-Ethyl-4-fluoro-1-nitrobenzene 97%

Supplier: Ambeed
Warning

A233833-100MG A233833-1G A233833-250MG
77098-304EA 51.99 USD
77098-304 77098-306 77098-308
2-Ethyl-4-fluoro-1-nitrobenzene 97%
2-Ethyl-4-fluoro-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.153 g/mol
MDL Number: MFCD16658616
CAS Number: 1089279-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR