(4-Chloro-3-(4-ethoxybenzyl)phenyl)((3aS,5R,6S,6aS)-6-hydroxy-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-5-yl)methanone 97%

Supplier: Ambeed
Warning

A233196-10G A233196-1G A233196-250MG A233196-5G
77098-288EA 1192.46 USD
77098-288 77098-290 77098-292 77098-294
(4-Chloro-3-(4-ethoxybenzyl)phenyl)((3aS,5R,6S,6aS)-6-hydroxy-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-5-yl)methanone 97%
(4-Chloro-3-(4-ethoxybenzyl)phenyl)((3aS,5R,6S,6aS)-6-hydroxy-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-5-yl)methanone

Precaución: for Research Use Only

Formula: C₂₃H₂₅ClO₆
MW: 432.89 g/mol
MDL Number: MFCD25977360
CAS Number: 1103738-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR