(R)-1-(3-Bromophenyl)ethanamine 97%

Supplier: Ambeed
Danger

A233062-10G A233062-1G A233062-25G A233062-5G
77098-280EA 131 USD
77098-280 77098-282 77098-284 77098-286
(R)-1-(3-Bromophenyl)ethanamine 97%
(R)-1-(3-Bromophenyl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MDL Number: MFCD00671637
CAS Number: 176707-77-0
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR