2-(3,5-Dibromo-4-hydroxyphenyl)acetic acid 95%

Supplier: Ambeed
Warning

A232978-1G A232978-250MG
77098-276EA 94 USD
77098-276 77098-278
2-(3,5-Dibromo-4-hydroxyphenyl)acetic acid 95%
2-(3,5-Dibromo-4-hydroxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₃
MDL Number: MFCD00544484
CAS Number: 24744-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR