3-Bromo-6-methoxy-2-phenyl-4H-chromen-4-one 95%

Supplier: Ambeed
Warning

A232870-100MG A232870-1G A232870-250MG
77098-270EA 23.34 USD
77098-270 77098-272 77098-274
3-Bromo-6-methoxy-2-phenyl-4H-chromen-4-one 95%
3-Bromo-6-methoxy-2-phenyl-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₆H₁₁BrO₃
MW: 331.16 g/mol
MDL Number: MFCD24386866
CAS Number: 215999-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR