Ethyl butyrylacetate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl Butyrylacetate, Ethyl 3-oxohexanoate

A232671-1000G A232671-100G A232671-25G A232671-500G A232671-5G
77098-260EA 257 USD
77098-260 77098-262 77098-264 77098-266 77098-268
Ethyl butyrylacetate 95%
Ethyl butyrylacetate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MDL Number: MFCD00009401
CAS Number: 3249-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR