Piperazine-1,4-diylbis((4-chlorophenyl)methanone) 95%

Supplier: Ambeed
Warning

A232066-1G A232066-5G
77098-246EA 5.31 USD
77098-246 77098-248
Piperazine-1,4-diylbis((4-chlorophenyl)methanone) 95%
Piperazine-1,4-diylbis((4-chlorophenyl)methanone)

Precaución: for Research Use Only

Formula: C₁₈H₁₆Cl₂N₂O₂
MW: 363.24 g/mol
MDL Number: MFCD00390326
CAS Number: 107785-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR