N-Cbz-N-methyl-L-leucine 98%

Supplier: Ambeed
Warning

Synonyms: Z-N-Me-Leu-OH

A231569-1G A231569-250MG A231569-25G A231569-5G
77098-230EA 12 USD
77098-230 77098-232 77098-234 77098-236
N-Cbz-N-methyl-L-leucine 98%
N-Cbz-N-methyl-L-leucine

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD00038521
CAS Number: 33099-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR