H-Leu-pNA·HCl 97%

Supplier: Ambeed
Warning

Synonyms: L-Leucyl-4-nitroanilide HCl, (S)-2-Amino-4-methyl-N-(4-nitrophenyl)pentanamide hydrochloride

A229813-1G A229813-5G
77098-202EA 38 USD
77098-202 77900-965
H-Leu-pNA·HCl 97%
H-Leu-pNA·HCl

Precaución: for Research Use Only

Formula: C₁₂H₁₇N₃O₃·HCl
MW: 287.7 g/mol
MDL Number: MFCD00058248
CAS Number: 16010-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR