3-Bromobenzoyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: m-Bromobenzoyl chloride, 3-Bromobenzoylchloride

A228929-25G A228929-5G
77098-188EA 34 USD
77098-188 77098-190
3-Bromobenzoyl chloride 98%
3-Bromobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇H₄BrClO
MW: 219.46 g/mol
MDL Number: MFCD00000669
CAS Number: 1711-09-7
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR