Benzimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 1H-Benzo[d]imidazole, 1H-Benzimidazole

A228912-1000G A228912-100G A228912-10G A228912-25G A228912-500G
77098-178EA 89.12 USD
77098-178 77098-180 77098-182 77098-184 77098-186
Benzimidazole 97%
Benzimidazole

Precaución: for Research Use Only

Formula: C₇H₆N₂
MW: 118.14 g/mol
MDL Number: MFCD00005585
CAS Number: 51-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR