Pinometostat (EPZ-5676) 98%

Supplier: Ambeed
Warning

A228248-10MG A228248-1MG A228248-2MG A228248-5MG
77098-162EA 74 USD
77098-162 77098-164 77098-166 77098-168
Pinometostat (EPZ-5676) 98%
Pinometostat (EPZ-5676)

Precaución: for Research Use Only

Formula: C₃₀H₄₂N₈O₃
MDL Number: MFCD24849416
CAS Number: 1380288-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR