2-(Difluoromethoxy)aniline 95%

Supplier: Ambeed
Warning

A228203-1G A228203-5G
77098-158EA 8 USD
77098-158 77098-160
2-(Difluoromethoxy)aniline 95%
2-(Difluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₇F₂NO
MDL Number: MFCD00190104
CAS Number: 22236-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR