4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole 97%

Supplier: Ambeed
Warning

A227476-100MG A227476-1G A227476-250MG A227476-5G
77098-144EA 20.16 USD
77098-144 77098-146 77098-148 77098-150
4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole 97%
4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₆HBr₂FN₂S
MW: 311.95 g/mol
MDL Number: MFCD26939241
CAS Number: 1347736-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR