D-(+)-Cystine 98%

Supplier: Ambeed
Warning

A227300-100G A227300-1G A227300-250MG A227300-25G A227300-5G
77098-134EA 91 USD
77098-134 77098-136 77098-138 77098-140 77098-142
D-(+)-Cystine 98%
D-(+)-Cystine

Precaución: for Research Use Only

Formula: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MDL Number: MFCD00002610
CAS Number: 349-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR