4-Bromo-1-methylimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1-methyl-1H-imidazole

A227136-1G A227136-250MG A227136-25G A227136-5G
77098-128EA 40.32 USD
77098-128 77098-130 77098-132 77568-200
4-Bromo-1-methylimidazole 98%
4-Bromo-1-methylimidazole

Precaución: For research use only.

Formula: C₄H₅BrN₂
MW: 161 g/mol
MDL Number: MFCD01320501
CAS Number: 25676-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR