(R)-3-Aminobutyric acid 97%

Supplier: Ambeed
Warning

A226851-100G A226851-10G A226851-1G A226851-25G A226851-5G A226851-500G
77098-114EA 17 USD
77098-114 77098-116 77098-118 77098-120 77098-122 77612-208
(R)-3-Aminobutyric acid 97%
(R)-3-Aminobutyric acid

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MDL Number: MFCD00211284
CAS Number: 3775-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR