1-Bromo-3-(butylsulfinyl)benzene 98%

Supplier: Ambeed
Warning

A226002-100MG A226002-1G A226002-250MG A226002-5G
77098-094EA 20 USD
77098-094 77098-096 77098-098 77098-100
1-Bromo-3-(butylsulfinyl)benzene 98%
1-Bromo-3-(butylsulfinyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrOS
MDL Number: MFCD20441854
CAS Number: 1215077-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR