α,α,α-Trifluoro-3,4-diaminotoluene 98%

Supplier: Ambeed
Warning

A225887-10G A225887-1G A225887-25G A225887-5G A225887-100G
77098-086EA 33 USD
77098-086 77098-088 77098-090 77098-092 77612-204
α,α,α-Trifluoro-3,4-diaminotoluene 98%
α,α,α-Trifluoro-3,4-diaminotoluene

Precaución: for Research Use Only

Formula: C₇H₇F₃N₂
MDL Number: MFCD00042456
CAS Number: 368-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR