tert-Butyl (3-((tert-butyldimethylsilyl)oxy)-2-oxopropyl)carbamate 95%

Supplier: Ambeed
Warning

A225769-100MG A225769-250MG A225769-5G
77098-080EA 17 USD
77098-080 77098-082 77098-084
tert-Butyl (3-((tert-butyldimethylsilyl)oxy)-2-oxopropyl)carbamate 95%
tert-Butyl (3-((tert-butyldimethylsilyl)oxy)-2-oxopropyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₉NO₄Si
MDL Number: MFCD24466259
CAS Number: 184429-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR