OICR-9429 97%

Supplier: Ambeed
Warning

Synonyms: N-[2-(4-Methylpiperazin-1-yl)-5-[3-(morpholin-4- ylmethyl)phenyl]phenyl]-6-oxo-4-(trifluoromethyl)-1,6-dihydropyridine-3-carboxamide, N-(4-(4-Methylpiperazin-1-yl)-3'-(morpholinomethyl)-[1,1'-biphenyl]-3-yl)-6-oxo-4-(trifluoromethyl)-1,6-dihydropyridine-3-carboxamide

A225196-10MG A225196-1MG A225196-25MG A225196-5MG A225196-50MG
77098-060EA 79.57 USD
77098-060 77098-062 77098-064 77098-066 77685-614
OICR-9429 97%
OICR-9429

Precaución: for Research Use Only

Formula: C₂₉H₃₂F₃N₅O₃
MW: 555.59 g/mol
MDL Number: MFCD28411621
CAS Number: 1801787-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR