6-Bromo-7-fluoro-1H-indazole 98%

Supplier: Ambeed
Warning

A225123-10G A225123-250MG A225123-5G A225123-100MG A225123-1G
77098-056EA 590 USD
77098-056 77098-058 77685-612 77901-805 77901-277
6-Bromo-7-fluoro-1H-indazole 98%
6-Bromo-7-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MDL Number: MFCD23704110
CAS Number: 1427396-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR