3-Fluoro-4-(piperazin-1-yl)benzonitrile 95%

Supplier: Ambeed
Warning

A225013-10G A225013-1G A225013-25G A225013-5G
77098-042EA 55 USD
77098-042 77098-044 77098-046 77098-048
3-Fluoro-4-(piperazin-1-yl)benzonitrile 95%
3-Fluoro-4-(piperazin-1-yl)benzonitrile

Precaución: for Research Use Only

Formula: C11H12FN3
MDL Number: MFCD03043990
CAS Number: 182181-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR