α-Bromo-m-toluonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Bromomethyl)benzonitrile, m-(Bromomethyl)benzonitrile

A224519-100G A224519-10G A224519-25G A224519-5G
77098-022EA 106 USD
77098-022 77098-024 77098-026 77098-028 77098-030
α-Bromo-m-toluonitrile 98%
α-Bromo-m-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
MDL Number: MFCD00001809
CAS Number: 28188-41-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR