5-Acetyl-2H-pyrazole-3-carboxylic acid ethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-acetyl-1H-pyrazole-5-carboxylate

A223505-100MG A223505-1G A223505-250MG
77098-006EA 28.65 USD
77098-006 77098-008 77098-010
5-Acetyl-2H-pyrazole-3-carboxylic acid ethyl ester 98%
5-Acetyl-2H-pyrazole-3-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₃
MW: 182.177 g/mol
MDL Number: MFCD21362351
CAS Number: 37622-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR