5-Amino-3-fluorobenzonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-5-fluorobenzonitrile, 3-Cyano-5-fluoroaniline

A222960-10G A222960-1G A222960-25G A222960-5G
77097-990EA 95.49 USD
77097-990 77097-992 77097-994 77097-996
5-Amino-3-fluorobenzonitrile 95%
5-Amino-3-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MW: 136.13 g/mol
MDL Number: MFCD05864357
CAS Number: 210992-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR