3-Chloropyridine-4-boronic acid, neopentyl glycol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Chloropyridin-4-yl)-5,5-dimethyl-1,3,2-dioxaborinane, 3-Chloro-4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine

A222170-100MG A222170-1G A222170-250MG
77097-982EA 11 USD
77097-982 77097-984 77097-986
3-Chloropyridine-4-boronic acid, neopentyl glycol ester 98%
3-Chloropyridine-4-boronic acid, neopentyl glycol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₃BClNO₂
MDL Number: MFCD06796314
CAS Number: 915070-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR